Learn More
Medchemexpress LLC Rebaudioside M | 1220616-44-3 | 10 MG
SDP
Catalog No. 5000417330
Encompass_Preferred
Rebaudioside M is a non-calorie sweetener isolated from *Stevia rebaudiana*. It is a glycoside of the *ent*-kaurene diterpenoid aglycone, over 350 times sweeter than sucrose, and suitable for use in food and beverages. This compound exhibits antidiabetic, antihypertension, anti-inflammatory, antioxidant, anticaries, and anticancer benefits. It is intended for research use only.
  • Non-calorie sweetener extracted from *Stevia rebaudiana*
  • Over 350 times sweeter than sucrose
  • Applicable in food and beverages
  • Demonstrates antidiabetic, antihypertension, anti-inflammatory, and antioxidant benefits
  • Supports β-cell activity and insulin secretion
  • Inhibits PI3K/Akt and Wnt/catenin signaling pathways
  • Helps lower blood glucose and blood pressure
Catalog No. 50-004-17330 Supplier Medchemexpress LLC Supplier No. HYN683310MG
Rebaudioside M is a non-calorie sweetener isolated from *Stevia rebaudiana*. It is a glycoside of the *ent*-kaurene diterpenoid aglycone, over 350 times sweeter than sucrose, and suitable for use in food and beverages. This compound exhibits antidiabetic, antihypertension, anti-inflammatory, antioxidant, anticaries, and anticancer benefits. It is intended for research use only.
  • Non-calorie sweetener extracted from *Stevia rebaudiana*
  • Over 350 times sweeter than sucrose
  • Applicable in food and beverages
  • Demonstrates antidiabetic, antihypertension, anti-inflammatory, and antioxidant benefits
  • Supports β-cell activity and insulin secretion
  • Inhibits PI3K/Akt and Wnt/catenin signaling pathways
  • Helps lower blood glucose and blood pressure