Learn More
Medchemexpress LLC Rebaudioside M (Standard) | 1220616-44-3 | 100 MG
SDP
Catalog No. 5000421011
Encompass_Preferred
Rebaudioside M (Standard) is the analytical standard of Rebaudioside M. This product is intended for research and analytical applications. It is a glycoside of the ent-kaurene diterpenoid aglycone and a non-calorie sweetener isolated from Stevia rebaudiana, being more than 350 times sweeter than sucrose.
  • Intended for research and analytical applications
  • Non-calorie sweetener
  • Isolated from Stevia rebaudiana
  • More than 350 times sweeter than sucrose
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-21011 Supplier Medchemexpress LLC Supplier No. HYN6833R100MG
Rebaudioside M (Standard) is the analytical standard of Rebaudioside M. This product is intended for research and analytical applications. It is a glycoside of the ent-kaurene diterpenoid aglycone and a non-calorie sweetener isolated from Stevia rebaudiana, being more than 350 times sweeter than sucrose.
  • Intended for research and analytical applications
  • Non-calorie sweetener
  • Isolated from Stevia rebaudiana
  • More than 350 times sweeter than sucrose
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS experiments