Learn More
Medchemexpress LLC Rebaudioside M (Standard) | 1220616-44-3 | 5 MG
SDP
Catalog No. 5000418519
Encompass_Preferred
Rebaudioside M (Standard) is an analytical standard for Rebaudioside M, a non-calorie sweetener isolated from *Stevia rebaudiana*. This glycoside of the ent-kaurene diterpenoid aglycone is more than 350 times sweeter than sucrose. It is intended for research and analytical applications.
  • Serves as an analytical standard and reference for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-18519 Supplier Medchemexpress LLC Supplier No. HYN6833R5MG
Rebaudioside M (Standard) is an analytical standard for Rebaudioside M, a non-calorie sweetener isolated from *Stevia rebaudiana*. This glycoside of the ent-kaurene diterpenoid aglycone is more than 350 times sweeter than sucrose. It is intended for research and analytical applications.
  • Serves as an analytical standard and reference for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS