Learn More
Medchemexpress LLC Rebaudioside M (standard) | 1220616-44-3 | 50 MG
SDP
Catalog No. 5000419489
Encompass_Preferred
Rebaudioside M (Standard) is the analytical standard of Rebaudioside M. It is a glycoside of the ent-kaurene diterpenoid aglycone, a non-calorie sweetener isolated from Stevia rebaudiana, and is more than 350 times sweeter than sucrose. This product is intended for research and analytical applications.
  • Analytical standard for Rebaudioside M
  • Intended for research and analytical applications
  • Glycoside of ent-kaurene diterpenoid aglycone
  • Non-calorie sweetener isolated from Stevia rebaudiana
  • More than 350 times sweeter than sucrose
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-19489 Supplier Medchemexpress LLC Supplier No. HYN6833R50MG
Rebaudioside M (Standard) is the analytical standard of Rebaudioside M. It is a glycoside of the ent-kaurene diterpenoid aglycone, a non-calorie sweetener isolated from Stevia rebaudiana, and is more than 350 times sweeter than sucrose. This product is intended for research and analytical applications.
  • Analytical standard for Rebaudioside M
  • Intended for research and analytical applications
  • Glycoside of ent-kaurene diterpenoid aglycone
  • Non-calorie sweetener isolated from Stevia rebaudiana
  • More than 350 times sweeter than sucrose
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS