Learn More
TARGETMOL CHEMICALS INC Rebaudioside N 1MG
SDP
Catalog No. 502880299
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Rebaudioside N is a steviol glycoside isolated from the leaves of Stevia rebaudiana and is used as a food additive. Purity 98.26%
Catalog No. 50-288-0299 Supplier TARGETMOL CHEMICALS INC Supplier No. TN21381MG
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Rebaudioside N is a steviol glycoside isolated from the leaves of Stevia rebaudiana and is used as a food additive. Purity 98.26%