Learn More
Lonza Walkersville REGM Renal Epithelial Cell Growth Medium BulletKit
Catalog No. NC9556824
Encompass
REGM Renal Epithelial Cell Growth Medium is formulated to maintain and expand most renal epithelial cells. REGM contains 0.5% serum.Select References: Diabetic Nephropathy is a chronic, progressive renal disease/diabetic kidney disease that develops in approximately one third of people with diabetes.In a study, Lonza’s human proximal tubular epithelial cells were cultured in respective media (REGM).Content:1 x REBM Basal Medium (CC-3191), 500 mL1 x REGM Singlequots Supplement Pack (CC-4127) containing:1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 2.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Triiodothyronine, 0.50 mL1 x Amber Vial with Epinephrine, 0.50 mL
Catalog No. NC9556824 Supplier Lonza Walkersville Supplier No. CC3190
REGM Renal Epithelial Cell Growth Medium is formulated to maintain and expand most renal epithelial cells. REGM contains 0.5% serum.Select References: Diabetic Nephropathy is a chronic, progressive renal disease/diabetic kidney disease that develops in approximately one third of people with diabetes.In a study, Lonza’s human proximal tubular epithelial cells were cultured in respective media (REGM).Content:1 x REBM Basal Medium (CC-3191), 500 mL1 x REGM Singlequots Supplement Pack (CC-4127) containing:1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 2.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Triiodothyronine, 0.50 mL1 x Amber Vial with Epinephrine, 0.50 mL