Learn More
Medchemexpress LLC Reboxetine-d5 (mesylate) | 1285918-53-7 | 95.0% | C20H22D5NO6S | 2.5 MG
SDP
Catalog No. 5000355452
Encompass_Preferred
Reboxetine-d5 mesylate is the deuterium labeled Reboxetine mesylatea. It is an orally active, potent, and selective noradrenaline uptake blocker, useful for antidepressant research. This compound serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, with deuteration potentially affecting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Orally active, potent, and selective noradrenaline uptake blocker
  • Useful for antidepressant research
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-55452 Supplier Medchemexpress LLC Supplier No. HY14560S2.5MG
Reboxetine-d5 mesylate is the deuterium labeled Reboxetine mesylatea. It is an orally active, potent, and selective noradrenaline uptake blocker, useful for antidepressant research. This compound serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, with deuteration potentially affecting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Orally active, potent, and selective noradrenaline uptake blocker
  • Useful for antidepressant research
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles