Learn More
Apexbio Technology LLC Reboxetine mesylate, 10mg. Cas: 98769-84-7 MFCD: MFCD02184016. Potent and selective inhibitor of noradrenalin uptake
SDP
Supplier:  Apexbio Technology LLC B212410
Encompass_Preferred
Reboxetine is a norepinephrine reuptake inhibitor with Ki of 8.2 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5310