Learn More
Apexbio Technology LLC Reboxetine mesylate 98769-84-7 50mg
SDP
Catalog No. 5000063460
Encompass_Preferred
Reboxetine mesylate (CAS 98769-84-7) is a selective inhibitor targeting norepinephrine reuptake It exhibits high affinity towards the norepinephrine transporter with a reported K i of approximately 8 2 nM thereby increasing synaptic norepinephrine concentrations Due to its selective modulatory action on noradrenergic pathways reboxetine mesylate is extensively utilized in neuroscience research investigating depressive disorders cognitive functions and associated biochemical mechanisms
Catalog No. 50-000-63460 Supplier Apexbio Technology LLC Supplier No. B212450
Reboxetine mesylate (CAS 98769-84-7) is a selective inhibitor targeting norepinephrine reuptake It exhibits high affinity towards the norepinephrine transporter with a reported K i of approximately 8 2 nM thereby increasing synaptic norepinephrine concentrations Due to its selective modulatory action on noradrenergic pathways reboxetine mesylate is extensively utilized in neuroscience research investigating depressive disorders cognitive functions and associated biochemical mechanisms