Learn More
Medchemexpress LLC Reboxetine mesylate | 98769-84-7 | MFCD02184016 | 99.7% | 409.50 g·mol⁻1 | C20H27NO6S | 5 MG
SDP
Catalog No. 5000221474
Encompass_Preferred
Reboxetine mesylate is the methanesulfonate salt of reboxetine, a selective noradrenaline reuptake inhibitor used as a research compound to study norepinephrine transporter function and neuropharmacology. It is supplied as a high-purity research chemical for in vitro and in vivo experimental applications; CAS 98769-84-7, molecular weight 409.50 g·mol⁻1.
  • Selective noradrenaline reuptake inhibitor activity for transporter studies.
  • Suitable for in vitro and in vivo research applications.
  • High purity suitable for analytical and pharmacological assays.
  • Available in small milligram quantities for laboratory use.
  • Provided with referenced product documentation and datasheet.
Catalog No. 50-002-21474 Supplier Medchemexpress LLC Supplier No. HY14560C5MG
Add to Cart
Add to Cart
Reboxetine mesylate is the methanesulfonate salt of reboxetine, a selective noradrenaline reuptake inhibitor used as a research compound to study norepinephrine transporter function and neuropharmacology. It is supplied as a high-purity research chemical for in vitro and in vivo experimental applications; CAS 98769-84-7, molecular weight 409.50 g·mol⁻1.
  • Selective noradrenaline reuptake inhibitor activity for transporter studies.
  • Suitable for in vitro and in vivo research applications.
  • High purity suitable for analytical and pharmacological assays.
  • Available in small milligram quantities for laboratory use.
  • Provided with referenced product documentation and datasheet.