Learn More
Abcam Rebo x etine mesylate, Noradrenaline reuptake inhibitor, 50MG
Catalog No. 502584675
Encompass_Preferred
MW 409.5 Da, Purity >99%. Potent, selective noradrenaline reuptake inhibitor, with high selectivity over dopamine and 5-HT transporters. Antidepressant in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4675 Supplier Abcam Supplier No. AB12015750MG
MW 409.5 Da, Purity >99%. Potent, selective noradrenaline reuptake inhibitor, with high selectivity over dopamine and 5-HT transporters. Antidepressant in vivo.
The product is subject to the following: Abcam Restricted Use Statement