Learn More
Cayman Chemical ReboxetInmesylate 10mg
SDP
Catalog No. 502972846
Encompass_Preferred
A selective norepinephrine reuptake inhibitor (IC50s = 8.2 nM) that displays greater than 1,000-fold affinity for binding rat norepinephrine receptors compared to serotonin, histamine, acetylcholine, or dopamine receptors (Kis = 1.1, 129, 1,400, 3,900, and >10,000 nM, respectively)
Catalog No. 50-297-2846 Supplier Cayman Chemical Supplier No. 1503810 MG
A selective norepinephrine reuptake inhibitor (IC50s = 8.2 nM) that displays greater than 1,000-fold affinity for binding rat norepinephrine receptors compared to serotonin, histamine, acetylcholine, or dopamine receptors (Kis = 1.1, 129, 1,400, 3,900, and >10,000 nM, respectively)