Learn More
Cayman Chemical Reboxetine (mesylate), Purity: ≥98%, CAS: 98769-84-7, 50 mg
SDP
Catalog No. 502970618
Encompass_Preferred
A selective norepinephrine reuptake inhibitor (IC50s = 8.2 nM) that displays greater than 1,000-fold affinity for binding rat norepinephrine receptors compared to serotonin, histamine, acetylcholine, or dopamine receptors (Kis = 1.1, 129, 1,400, 3,900, and >10,000 nM, respectively)
Catalog No. 50-297-0618 Supplier Cayman Chemical Supplier No. 1503850 MG
A selective norepinephrine reuptake inhibitor (IC50s = 8.2 nM) that displays greater than 1,000-fold affinity for binding rat norepinephrine receptors compared to serotonin, histamine, acetylcholine, or dopamine receptors (Kis = 1.1, 129, 1,400, 3,900, and >10,000 nM, respectively)