Learn More
SOMNI Scientific™ Rebreathing Small Animal Anesthesia Machine
SDP
Catalog No. 04355313 Shop All Somni Scientific Products
Change view
Click to view available options
Animal:
Large Animal
Catalog No. Animal
04-355-313 Large Animal
1 options
Catalog No. 04-355-313 Supplier SOMNI Scientific™ Supplier No. RM17013
Add to Cart
Add to Cart
Includes Adult and Ped F-circuit and 2 L bag
  • High quality limited maintenance design
  • Pressure manometer center mounted for immediate visibility
  • Toggle switch: rebreathing to nonrebreathing, circuits stay in place
  • H-frame with shelf provides support and stability
  • 3 in. wheels for maneuverability
  • Popoff valve releases at 2 cm H2O while maintaining passive volume
  • Supports oxygen concentrator
  • Small footprint, easy assembly
  • Powder coated and anodized frame for easy cleaning
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Animal Large Animal
Type Anesthesia Machine