Learn More
Waters Corp Rebuild Kit, Divert Valve, ACQUITY QDa System
Catalog No. 502391423 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit is designed for a divert valve, an optional accessory for the ACQUITY QDa Detector, ensuring its continued optimal performance. The divert valve plays a crucial role in managing sample flow within the system, allowing for precise handling of various components.
  • Supports functions such as diverting high concentration components to waste.
  • Facilitates the diversion of matrix components to waste.
  • Enables manual loop injections of samples.
  • Operates in two primary modes for flexible sample management.
  • Designed for mass spectrometry systems.
  • Recommended for annual replacement to maintain performance.
Catalog No. 50-239-1423 Supplier Waters Corp Supplier No. 700011201
This rebuild kit is designed for a divert valve, an optional accessory for the ACQUITY QDa Detector, ensuring its continued optimal performance. The divert valve plays a crucial role in managing sample flow within the system, allowing for precise handling of various components.
  • Supports functions such as diverting high concentration components to waste.
  • Facilitates the diversion of matrix components to waste.
  • Enables manual loop injections of samples.
  • Operates in two primary modes for flexible sample management.
  • Designed for mass spectrometry systems.
  • Recommended for annual replacement to maintain performance.