Learn More
Grainger Danfoss Rebuild Kit, For EVR 20 and 22 Solenoid Valves, 1 ea
Catalog No. 502761928
Encompass_Preferred
This service kit is designed for the maintenance and repair of solenoid valves. It is compatible with specific models to ensure proper operation and performance in air conditioning and commercial refrigeration systems.
  • Designed for use with EVR 20 and EVR 22 solenoid valves
  • Includes components for a.c. service
  • Compatible with manufacturer model number 032F1176
  • Supplied as a single unit
Catalog No. 50-276-1928 Supplier Grainger Supplier No. 161J35
This service kit is designed for the maintenance and repair of solenoid valves. It is compatible with specific models to ensure proper operation and performance in air conditioning and commercial refrigeration systems.
  • Designed for use with EVR 20 and EVR 22 solenoid valves
  • Includes components for a.c. service
  • Compatible with manufacturer model number 032F1176
  • Supplied as a single unit