Learn More
Chemglass Life Sciences Rebuild Kit for Agitator Assemblies, 8mm Shaft Size
SDP
Catalog No. 501294506
Encompass
Rebuild Kit for Agitator Assemblies, 8mm Shaft Size.Kits of o-rings, seals and bearings to rebuild agitator assemblies.CLS-1380-05PM fits agitator assemblies: CLS-1380-05, CLS-1383-04, CLS-1385-35, Shaft Size 8mm.
Catalog No. 50-129-4506 Supplier Chemglass Life Sciences Supplier No. CLS138005PM
Rebuild Kit for Agitator Assemblies, 8mm Shaft Size.Kits of o-rings, seals and bearings to rebuild agitator assemblies.CLS-1380-05PM fits agitator assemblies: CLS-1380-05, CLS-1383-04, CLS-1385-35, Shaft Size 8mm.