Learn More
Electron Microscopy Sciences EMS Tungsten Loop Filament Rebuilt, ISI, ABT, Topcon
SDP
Catalog No. 5029075
Encompass_Preferred
Filament rebuilding includes reconditioning of the base; cleaning and/or replacement of necessary parts; Installment of new filament tip; Pre-centering; Vacuum normalizing; and Microscopic inspection.
Use: ISI Instruments, ABT, Topcon
Catalog No. 50-290-75 Supplier Electron Microscopy Sciences Supplier No. 81038R
Filament rebuilding includes reconditioning of the base; cleaning and/or replacement of necessary parts; Installment of new filament tip; Pre-centering; Vacuum normalizing; and Microscopic inspection.
Use: ISI Instruments, ABT, Topcon