Learn More
TARGETMOL CHEMICALS INC REC 15/2615 DIHYDROCHLOR 5MG
SDP
Catalog No. 502580887
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Rec 15/2615 dihydrochloride is an antagonist of (alpha)1B-adrenergic receptor. purity: 99%
Catalog No. 50-258-0887 Supplier TARGETMOL CHEMICALS INC Supplier No. T232305MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Rec 15/2615 dihydrochloride is an antagonist of (alpha)1B-adrenergic receptor. purity: 99%