Learn More
Biotang Inc Avidin,Activity 11-15 units/mg protein, 1 unit binds 1ug biotin. Application: Avidin may be used to visualize biotin conjugated molecules in ELISA, blotting and histological techniques
Supplier:  Biotang Inc BTPRO500
Encompass_Preferred
Avidin,Activity 11-15 units/mg protein, 1 unit binds 1ug biotin. Application: Avidin may be used to visualize biotin conjugated molecules in ELISA, blotting and histological techniques
Catalog No. 50-148-925