Learn More
Biotang Inc Alkaline Phosphatase Bovine Intestinal, Alkaline Phosphatase Bovine, Alkaline Phosphatase is Dimeric protein having a molecular weight of 140 kDa, Mg++ stimulates the catalysis.
Catalog No. 50148920 Shop All BIOTANG Inc Products
Encompass_Preferred
Alkaline Phosphatase Bovine Intestinal, Alkaline Phosphatase Bovine, Alkaline Phosphatase is Dimeric protein having a molecular weight of 140 kDa, Mg++ stimulates the catalysis.
Catalog No. 50-148-920 Supplier Biotang Inc Supplier No. BTENZ322
Alkaline Phosphatase Bovine Intestinal, Alkaline Phosphatase Bovine, Alkaline Phosphatase is Dimeric protein having a molecular weight of 140 kDa, Mg++ stimulates the catalysis.