Learn More
Biotang Inc Aprotinin, Activity 6,462 KIU (Kallikrein Inactivator Units) per mg, 4.97 TIU/mg.Greater than 98.0% as determined by(a) Analysis by RP-HPLC.
Supplier:  Biotang Inc BTPRO285
Encompass_Preferred
Aprotinin, Activity 6,462 KIU (Kallikrein Inactivator Units) per mg, 4.97 TIU/mg.Greater than 98.0% as determined by(a) Analysis by RP-HPLC.
Catalog No. 50-148-931