Learn More
Active Motif REC.BRDT(257-382),GST-TAG,1MG
SDP
Supplier:  Active Motif 31883
Encompass
The peptide corresponding to amino acids 257-382 that contains the second bromodomain sequences of BRDT (accession number NP_997072.1) was expressed in E. coli and contains an N-terminal GST-Tag with an observed molecular weight of 42.8 kDa. The recombinant protein is >90% pure by SDS-PAGE.
Catalog No. NC1930406
Edge