Learn More
Biotang Inc Cystatin-C Canine Recombinant, Cystatin C k9, Cystatin-C Protein is a 14.85 kDa protein containing 133 amino acids fused to a 8 aa N-Terminal His-tag, Synonyms: Cystatin-C, Cystatin-3, Gamma-trace, Post-gamma-globulin, CST3, MGC117328
Supplier:  Biotang Inc BTPRO138
Encompass_Preferred
Cystatin-C Canine Recombinant, Cystatin C k9, Cystatin-C Protein is a 14.85 kDa protein containing 133 amino acids fused to a 8 aa N-Terminal His-tag, Synonyms: Cystatin-C, Cystatin-3, Gamma-trace, Post-gamma-globulin, CST3, MGC117328
Catalog No. 50-148-961