Learn More
Perkin Elmer US LLC Special Optical Glass Rectangular Macro Cell with PTFE Lid - 50 mm, Qty. 1
Catalog No. 509042612
Encompass_Preferred
The Special Optical Glass Macro Cell has a 10 mm path length, a cell volume of 3.5 mL, and a PTFE lid. Its outside dimensions are 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2
Catalog No. 50-904-2612 Supplier Perkin Elmer US LLC Supplier No. B0631005
The Special Optical Glass Macro Cell has a 10 mm path length, a cell volume of 3.5 mL, and a PTFE lid. Its outside dimensions are 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2