Learn More
Biomatik Corporation Recombinant Human Chemokine (C-X-C Motif) Ligand 14 (CXCL14)
Catalog No. NC2684287
Encompass_Preferred
Recombinant Human Chemokine (C-X-C Motif) Ligand 14 (CXCL14) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Chemokine (C-X-C Motif) Ligand 14 (CXCL14). Target Synonyms: BRAK; SCYB14; BRAK; NJAC; Bolekine; Kec; MIP2-G; MIP2g; BMAC; KS1; Breast And Kidney Expressed Chemokine; Small Inducible Cytokine Subfamily B(Cys-X-Cys)Member 14. Accession Number: O95715. Gene ID: 9547. Expression Region: Ser35~Glu111. Tag Info: N-terminal His and GST Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2684287 Supplier Biomatik Corporation Supplier No. RPU52406
Recombinant Human Chemokine (C-X-C Motif) Ligand 14 (CXCL14) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Chemokine (C-X-C Motif) Ligand 14 (CXCL14). Target Synonyms: BRAK; SCYB14; BRAK; NJAC; Bolekine; Kec; MIP2-G; MIP2g; BMAC; KS1; Breast And Kidney Expressed Chemokine; Small Inducible Cytokine Subfamily B(Cys-X-Cys)Member 14. Accession Number: O95715. Gene ID: 9547. Expression Region: Ser35~Glu111. Tag Info: N-terminal His and GST Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.