Learn More
Biotang Inc Ecotin E.Coli Recombinant, Ecotin produced in E.Coli is a single, non-glycosylated polypeptide chain containing 163 amino acids (21-162a.a.) and having a molecular mass of 18.3kDa.
Supplier:  Biotang Inc BTENZ058
Encompass_Preferred
Ecotin E.Coli Recombinant, Ecotin produced in E.Coli is a single, non-glycosylated polypeptide chain containing 163 amino acids (21-162a.a.) and having a molecular mass of 18.3kDa.
Catalog No. 50-148-999