Learn More
Abcam Recombinant E. coli RecQ DNA Helicase protein (Active), 20UG
Supplier:  Abcam AB6381320UG
Encompass_Preferred
Recombinant E. coli RecQ DNA Helicase protein (Active) is a Escherichia coli K-12 Full Length protein, in the 1 to 609 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: b3822, JW5855, recQ, ATP-dependent DNA helicase RecQ, DNA 3'-5' helicase RecQ
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1770129