Learn More
Abcam Recombinant EBV Nuclear Antigen/EBNA1 protein, 250UG
Supplier:  Abcam AB138345250UG
Encompass_Preferred
Recombinant EBV Nuclear Antigen/EBNA1 protein is a Epstein-Barr Virus (Strain B95-8) Full Length protein, in the 1 to 640 aa range with >95% purity and suitable for western blot, SDS-PAGE and ELISA.The predicted molecular weight of ab138345 protein is 63 kDa. - Save time and ensure accurate results- use our EBV Nuclear Antigen/EBNA1 protein as a control
Protein/ Peptide/ Enzyme Synonyms: BKRF1, EBNA1, Epstein-Barr nuclear antigen 1, EBNA-1, EBV nuclear antigen 1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2064161