Learn More
Bachem rec EGF (human)
SDP
Supplier:  Bachem 4030572.0500
Encompass_Preferred
rec EGF (human), H-Asn-Ser-Asp-Ser-Glu-Cys-Pro-Leu-Ser-His-Asp-Gly-Tyr-Cys-Leu-His-Asp-Gly-Val-Cys-Met-Tyr-Ile-Glu-Ala-Leu-Asp-Lys-Tyr-Ala-Cys-Asn-Cys-Val-Val-Gly-Tyr-Ile-Gly-Glu-Arg-Cys-Gln-Tyr-Arg-Asp-Leu-Lys-Trp-Trp-Glu-Leu-Arg-OH, 62253-63-8, C270H395N73O83S7, Mr 6216.01, 0.5mg. Nepidermin. approx. ED50 ≤ 0.1 ng/mL Human EGF (urogastrone, nepidermin) is a polypeptide hormone which was first isolated from human urine. It is a potent mitogen for a variety of cultured cells, e.g. fibroblasts. In vivo, urogastrone may have a role in the maintenance of gastrointestinal homeostasis. Furthermore, EGF is used in wound healing.
Catalog No. 50-260-155