Learn More
Biotang Inc Eledoisin, ledoisini-s molecular weight is 1188.4 having an amino acid sequence of Glp-Pro-Ser-Lys-Asp-Ala-Phe-Ile-Gly-Leu-Met-NH2 and a molecular formula of C54H85N13O15S.
Supplier:  Biotang Inc BTPRO281
Encompass_Preferred
Eledoisin, ledoisini-s molecular weight is 1188.4 having an amino acid sequence of Glp-Pro-Ser-Lys-Asp-Ala-Phe-Ile-Gly-Leu-Met-NH2 and a molecular formula of C54H85N13O15S.
Catalog No. 50-149-017