Learn More
Biotang Inc Exenatide, Exenatide is a single, non-glycosylated, peptide containing 39 amino acids and having a molecular mass of 4186.6 Dalton. Exenatide has the empirical formula C184H282N50O60S.
Catalog No. 50149019 Shop All BIOTANG Inc Products
Encompass_Preferred
Exenatide, Exenatide is a single, non-glycosylated, peptide containing 39 amino acids and having a molecular mass of 4186.6 Dalton. Exenatide has the empirical formula C184H282N50O60S.
Catalog No. 50-149-019 Supplier Biotang Inc Supplier No. BTHOR246
Exenatide, Exenatide is a single, non-glycosylated, peptide containing 39 amino acids and having a molecular mass of 4186.6 Dalton. Exenatide has the empirical formula C184H282N50O60S.