Learn More
Abcam Recombinant Human APT-2 protein, 100UG
Supplier:  Abcam AB105129100UG
Encompass_Preferred
Recombinant Human APT-2 protein is a Human Full Length protein, in the 1 to 231 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: APT2, LYPLA2, Acyl-protein thioesterase 2, APT-2, Lysophospholipase II, Palmitoyl-protein hydrolase, LPL-II, LysoPLA II
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2156893