Learn More
Abcam Recombinant Human Mesothelin protein, 50UG
Supplier:  Abcam AB20521750UG
Encompass_Preferred
Recombinant Human Mesothelin protein is a Human Full Length protein, in the 37 to 598 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: MPF, MSLN, Mesothelin, CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2000341