Learn More
Abcam Recombinant Human NLRP3 protein, 5UG
Supplier:  Abcam AB1980695UG
Encompass_Preferred
Recombinant Human NLRP3 protein is a Human Full Length protein, in the 2 to 1036 aa range, expressed in Baculovirus infected Sf9, with >=50% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: C1orf7, CIAS1, NALP3, PYPAF1, NLRP3, Angiotensin/vasopressin receptor AII/AVP-like, Caterpiller protein 1.1, Cold-induced autoinflammatory syndrome 1 protein, Cryopyrin, PYRIN-containing APAF1-like protein 1, CLR1.1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1842107