Learn More
Abcam Recombinant human PLA2G7 protein (Active), 20UG
Supplier:  Abcam AB27171520UG
Encompass_Preferred
Recombinant human PLA2G7 protein (Active) is a Human Full Length protein, in the 22 to 441 aa range, expressed in HEK 293, with >=91% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: PAFAH, PLA2G7, Platelet-activating factor acetylhydrolase, PAF acetylhydrolase, 1-alkyl-2-acetylglycerophosphocholine esterase, 2-acetyl-1-alkylglycerophosphocholine esterase, Group-VIIA phospholipase A2, LDL-associated phospholipase A2, PAF 2-acylhydrolase, gVIIA-PLA2, LDL-PLA(2)
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2116001