Learn More
Abcam Recombinant HPV33 L1 protein, 50UG
Supplier:  Abcam AB11988250UG
Encompass_Preferred
Recombinant HPV33 L1 protein is a human papillomavirus 33 Full Length protein, in the 1 to 499 aa range, expressed in Saccharomyces cerevisiae, with >90% purity and suitable for SDS-PAGE, EM, WB, ELISA.
Protein/ Peptide/ Enzyme Synonyms: Major capsid protein L1, L1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2137356