Learn More
Abcam Recombinant Human Axin 1 protein, 50UG
Supplier:  Abcam AB20409050UG
Encompass_Preferred
Recombinant Human Axin 1 protein is a Human Full Length protein, in the 1 to 826 aa range, expressed in Baculovirus infected Sf9, with >75% purity and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: AXIN, AXIN1, Axin-1, Axis inhibition protein 1, hAxin
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2291213