Learn More
Apexbio Technology LLC REC HU M-CSF HIS-FLAG-1mg
SDP
Catalog No. 5000897650
Encompass_Preferred
Recombinant Human M-CSF/CSF1 (His Flag) is a crucial cytokine involved in the regulation of immune system functions Originating from human sources it plays a significant role in hematopoietic stem cell differentiation and macrophage lineage fate by binding to the CSF1 receptor Its mechanism of action involves the activation of signaling pathways that promote cell proliferation and survival This cytokine demonstrates potent activity in the nanomolar to low micromolar range when assessed in vitro It is commonly applied in cell models such as macrophage colony assays and utilized in animal studies to investigate immune responses and bone metabolism Additionally it serves an essential role in drug screening processes aimed at developing therapies for immune-related conditions Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97650 Supplier Apexbio Technology LLC Supplier No. PH10231
Recombinant Human M-CSF/CSF1 (His Flag) is a crucial cytokine involved in the regulation of immune system functions Originating from human sources it plays a significant role in hematopoietic stem cell differentiation and macrophage lineage fate by binding to the CSF1 receptor Its mechanism of action involves the activation of signaling pathways that promote cell proliferation and survival This cytokine demonstrates potent activity in the nanomolar to low micromolar range when assessed in vitro It is commonly applied in cell models such as macrophage colony assays and utilized in animal studies to investigate immune responses and bone metabolism Additionally it serves an essential role in drug screening processes aimed at developing therapies for immune-related conditions Experimental concentrations typically depend on the specific research design