Learn More
Apexbio Technology LLC REC HU TPO HIS-FLAG-1mg
SDP
Catalog No. 5000897641
Encompass_Preferred
Recombinant Human Thrombopoietin/TPO (His Flag) is a biotechnologically produced protein used in the field of hematopoiesis research Originating from human sources it primarily promotes the proliferation and differentiation of megakaryocyte progenitor cells via binding to the c-Mpl receptor activating JAK-STAT pathways In vitro its activity is often observed in the nanomolar to low micromolar range making it a valuable tool in studying thrombopoiesis It is commonly applied in cell culture models animal studies and drug screening assays providing insights into platelet production and related disorders Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97641 Supplier Apexbio Technology LLC Supplier No. PH10201
Recombinant Human Thrombopoietin/TPO (His Flag) is a biotechnologically produced protein used in the field of hematopoiesis research Originating from human sources it primarily promotes the proliferation and differentiation of megakaryocyte progenitor cells via binding to the c-Mpl receptor activating JAK-STAT pathways In vitro its activity is often observed in the nanomolar to low micromolar range making it a valuable tool in studying thrombopoiesis It is commonly applied in cell culture models animal studies and drug screening assays providing insights into platelet production and related disorders Experimental concentrations typically depend on the specific research design