Learn More
Biotang Inc Beta 2 Microglobulin Human Recombinant, 200ug B2M Human Recombinant, B2 Microglobulin Human Recombinant produced in E.Coli is a non-glycosylated polypeptide chain having a molecular mass of 11.76 kDa.
Supplier:  Biotang Inc BTPRO553
Encompass_Preferred
Beta 2 Microglobulin Human Recombinant, 200ug B2M Human Recombinant, B2 Microglobulin Human Recombinant produced in E.Coli is a non-glycosylated polypeptide chain having a molecular mass of 11.76 kDa.
Catalog No. 50-149-236