Learn More
Biotang Inc B-type Natriuretic Peptide Human Recombinant, Synonyms NPPB, Natriuretic Peptide Precursor B, BNP, B-type Natriuretic Peptide. non-glycosylated, polypeptide chain containing 32 amino acids and having a molecular mass of 3464 Dalton.
Supplier:  Biotang Inc BTCYT327
Encompass_Preferred
B-type Natriuretic Peptide Human Recombinant, Synonyms NPPB, Natriuretic Peptide Precursor B, BNP, B-type Natriuretic Peptide. non-glycosylated, polypeptide chain containing 32 amino acids and having a molecular mass of 3464 Dalton.
Catalog No. 50-149-274