Learn More
Biotang Inc REC HUM BPGM 5UG
Supplier:  Biotang Inc BTENZ505
Encompass_Preferred
Human 2,3-Bisphosphoglycerate Mutase BPGM Human Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 267 amino acids (1-259 a.a.) and having a molecular mass of 31 kDa. The BPGM is fused to an 8 amino acid His Tag at C-Terminus and purified by proprietary chromatographic techniques. Greater than 95% as determined by SDS-PAGE.
Catalog No. 50-149-049