Learn More
Biotang Inc Buserelin, > 98%, Buserelin contains 9 amino acids Glu-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHEt and having a molecular weight of 1239.44 Dalton.
Catalog No. 50149277 Shop All BIOTANG Inc Products
Encompass_Preferred
Buserelin, > 98%, Buserelin contains 9 amino acids Glu-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHEt and having a molecular weight of 1239.44 Dalton.
Catalog No. 50-149-277 Supplier Biotang Inc Supplier No. BTHOR255
Buserelin, > 98%, Buserelin contains 9 amino acids Glu-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHEt and having a molecular weight of 1239.44 Dalton.