Learn More
Biotang Inc Follicle Stimulating Hormone Human Recombinant, FSH Human,The ED50 as determined by cAMP accumulation in human FSH Receptor transfected Chinese Hamster Ovary cells was found to be 80-450 pg/ml.
Supplier:  Biotang Inc BTHOR253
Encompass_Preferred
Follicle Stimulating Hormone Human Recombinant, FSH Human,The ED50 as determined by cAMP accumulation in human FSH Receptor transfected Chinese Hamster Ovary cells was found to be 80-450 pg/ml.
Catalog No. 50-149-616