Learn More
Biotang Inc Human Glucagon Like Peptide-1, GLP 1 Human,Glucagon Like Peptide-1 is a single, non-glycosylated, polypeptide chain containing 30 amino acids and having a molecular mass of 3298.7 Dalton.
Supplier:  Biotang Inc BTHOR284
Encompass_Preferred
Human Glucagon Like Peptide-1, GLP 1 Human,Glucagon Like Peptide-1 is a single, non-glycosylated, polypeptide chain containing 30 amino acids and having a molecular mass of 3298.7 Dalton.
Catalog No. 50-149-668