Learn More
Biotang Inc HIV-2 gp32 Recombinant, HIV-2 gp32, ontains the full-length sequence of HIV-2 envelope immunodominant regions gp32. The protein is fused with b-galactosidase (114 kDa) at N-terminus
Catalog No. 50151716 Shop All BIOTANG Inc Products
Encompass_Preferred
HIV-2 gp32 Recombinant, HIV-2 gp32, ontains the full-length sequence of HIV-2 envelope immunodominant regions gp32. The protein is fused with b-galactosidase (114 kDa) at N-terminus
Catalog No. 50-151-716 Supplier Biotang Inc Supplier No. BTHIV134
HIV-2 gp32 Recombinant, HIV-2 gp32, ontains the full-length sequence of HIV-2 envelope immunodominant regions gp32. The protein is fused with b-galactosidase (114 kDa) at N-terminus