Learn More
Biotang Inc Inhibin-Alpha A Chain Human Recombinant, containing 134 amino acids fragment (233-366) and having an amino-terminal hexahistidine tag, having a total molecular weight of 19.2 kDa.
Supplier:  Biotang Inc BTHOR293
Encompass_Preferred
Inhibin-Alpha A Chain Human Recombinant, containing 134 amino acids fragment (233-366) and having an amino-terminal hexahistidine tag, having a total molecular weight of 19.2 kDa.
Catalog No. 50-149-826