Learn More
Biotang Inc Myeloperoxidase Human, MPO Human, is a natural protein having a molecular mass of 150kDa containing 2 subunits each of a heavy chain with 64kDa and a light chain with 13kDa.isolated from human peripheral blood polymorphonuclear leukocytes.
Supplier:  Biotang Inc BTENZ074
Encompass_Preferred
Myeloperoxidase Human, MPO Human, is a natural protein having a molecular mass of 150kDa containing 2 subunits each of a heavy chain with 64kDa and a light chain with 13kDa.isolated from human peripheral blood polymorphonuclear leukocytes.
Catalog No. 50-150-093