Learn More
Biotang Inc Myoglobin His Human Recombinant, produced in E.Coli is a single, non-glycosylated polypeptide chain containing 174 amino acids (1-154a.a.) and having a molecular mass of 19.3kDa.fused to a 20 amino acid His-tag at N-terminus
Supplier:  Biotang Inc BTPRO071
Encompass_Preferred
Myoglobin His Human Recombinant, produced in E.Coli is a single, non-glycosylated polypeptide chain containing 174 amino acids (1-154a.a.) and having a molecular mass of 19.3kDa.fused to a 20 amino acid His-tag at N-terminus
Catalog No. 50-150-097